Learn More
Thermo Scientific™ FHZ 632 L External Dose Rate Probe
Provides external dose rates up to 10 Rh. For use with FH40 and FH40TG telescope pole.
Supplier: Thermo Scientific™ 4254056
This item is not returnable.
View return policy
Description
Specifications
36 keV to 1.3 MeV | |
Dose Rate Probe | |
FH40 and FH40TG Telescope Pole |