Learn More
PRO TEX INTERNATIONAL INC Face Shield Long 8x10 (12)
Supplier: PRO TEX INTERNATIONAL INC DFS8
PRO-TEX Clip-on Face Shields, Size Long 8x10, 12/Box, Clips on easily to your prescription glasses, safety glasses, or support frames. As a professional face shield, it is ideal for those in the medical or dental fields, including those who perform dialysis. When worn with a mask, they provide additional protection against infectious bodily spatter and spray.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.