Learn More
Hanna Pharmaceutical Supply Co., Inc. F/AIR CANISTER 200GM
Supplier: Hanna Pharmaceutical Supply Co., Inc. 80120
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The F/AIR anesthesia gas filter was specifically designed to remove waste anesthesia gases such as ISOFLURANCE, HALOTHANE, ENFLURANE, etc. from the environment. 8/BX
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable.
View return policy