Learn More
Restek™ EZ Twist Top Injection Port Kits
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Change inlet liners faster and easier, for Agilent 5890/6850/6890/7820 GCs
Supplier: Restek™ 22725
Description
- Gas lines are attached to shell weldment bottom to help eliminate broken gas lines
- Gas lines do not interfere with routine maintenance
- Weldment Removal Tool allows for quick removal of the hot weldment without fingers ever touching hot surfaces
- Weldment stays secure in the tool for reattachment
Specifications
Injection Port Kit | |
Split/Splitless | |
EZ Twist Top | |
Agilent 5890 |