Learn More
Medchemexpress LLC Everolimus Solution 10 mM * 1 mL in DMSO
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Medchemexpress LLC HY1021810MM1MLDMSO
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Everolimus (RAD001) is a Rapamycin (HY-10219) derivative and a potent, selective and orally active mTOR1 inhibitor. Everolimus binds to FKBP-12 to generate an immunosuppressive complex. Everolimus inhibits tumor cells proliferation and induces cell apoptosis and autophagy. Everolimus has potent immunosuppressive and anticancer activities.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.