Learn More
Electron Microscopy Sciences Electrically/Thermally Conductive Adhesive ETC Bond 598 50 GR
Supplier: Electron Microscopy Sciences 50380598
Electrically and thermally conductive adhesive, Nickel filled, One part system, Low viscosity, For applications up to 1000 deg F.