Learn More
Maine Molecular Quality Controls Inc ePlex BCID-FP Control M320 v1.1, 12 tubes x 50 uL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Maine Molecular Quality Controls Inc M320V1.1
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
The ePlex BCID-FP Control M320 v1.1 is intended for use as an external positive and negative quality control to monitor the performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection of Candida albicans, Candida auris, Candida dubliniensis, Candida famata, Candida glabrata, Candida guilliermondii, Candida kefyr, Candida krusei, Candida lusitaniae, Candida parapsilosis, Candida tropicalis, Cryptococcus gattii, Cryptococcus neoformans, Fusarium, and Rhodotorula on the ePlex Blood Culture Identification Fungal Pathogen (BCID-FP) Panel performed on the ePlex System (GenMark Diagnostics, Inc.). ePlex BCID-FP Control M320 v1.1 is composed of 3 controls, ePlex BCID-FP Positive A1, ePlex BCID-FP Positive B1, and ePlex BCID-FP Negative. ePlex BCID-FP Control M320 v1.1 is comprised of 12 tubes, 4 tubes of each positive control, A1 and B1, and 4 tubes of negative control, 50μL each.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.