Learn More
Bioassay Systems EnzyChrom™ Glutamine Assay Kit. For quantitative determination of glutamine and evaluation of drug effects on glutamine metabolism.
Supplier: Bioassay Systems EGLN100
For quantitative determination of glutamine and evaluation of drug effects on glutamine metabolism. Key Features: Sensitive and accurate. Use 20 µL sample. Linear detection range 0.023 - 2 mM glutamine in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, incubation for 40 min at room temperature, adding a stop reagent and reading the optical density. No 37°C heater is needed. High-throughput. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD565nm. Samples: Serum, plasma, urine, cell, tissue etc. Species: All. Procedure: Assay takes 40 min. Kit size: 100 tests. Detection limit: 23 µM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.