Learn More
Environmental Express™ Filters
Ideal for air particle monitoring and environmental testing
Supplier: Environmental Express™ F470100PP
This item is not returnable.
View return policy
Description
- Laser cutting ensures a clean, fiber-free filter edge
- Excellent temperature and chemical resistance
Specifications
47 mm |