Learn More
World Precision Instrument Endoscope Sterilization Baskets-WP-4047
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument WP4047
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
- Safely sterilize and store expensive endoscopes
- Silicone pads to protect endoscope during handling and storage
- Electropolished
- Burr free to prevent injury
- Locking clip
- Variety of sizes