Learn More
PREDICTIVE ONCOLOGY INC EndoPrep Kit
Supplier: PREDICTIVE ONCOLOGY INC EDP4001.01
EndoPrep™ is a sample treatment system to remove the inhibitory or enhancing effects of proteins, peptides or antibodies in endotoxin assays. This allows for more accurate endotoxin detection and quantitation. The EndoPrep™ system is compatible with both classical LAL and recombinant Factor C endotoxin detection assays.