Learn More
Thermo Scientific Enhanced Matrix Tolerance (EMT) Plasma Torch for the iCAP™ Series ICP-OES Systems
Supplier: Thermo Scientific BRE0012328
Enhanced Matrix Tolerance (EMT) Plasma Torch for the iCAP™ Series ICP-OES Systems, Enable simple, routine maintenance through the unique design of the Thermo Scientific™ Enhanced Matrix Tolerance (EMT) Plasma Torch for Thermo Scientific™ iCAP™ Series ICP-OES systems. The torch clips in and can be removed in just seconds.
This item is not returnable.
View return policy