Learn More
Electron Microscopy Sciences PolarStat™ Polarstat Embedding Medium 4 OZ
Supplier: Electron Microscopy Sciences 19222
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
An aqueous-based frozen section embedding medium designed to support tissue blocks in cryostat sectioning. Formulated to promote rapid freezing, enhanced sectioning and consistent results at a working temperature of -20 degree C.