Learn More
Electron Microscopy Sciences PolarStat™ Plus Embedding Medium
Supplier: Electron Microscopy Sciences 19221
This formulation is especially well suited for Mohs cryostat sectioning. It freezes at optimum cutting temperatures of -10 deg C and below. No residue is left on slides during the staining procedure, thereby eliminating undesirable background staining.