Learn More
Biomatik Corporation Phospho-p38 MAPK (Thr179+Tyr181) Colorimetric Cell-Based ELISA Kit
Supplier: Biomatik Corporation EKA51238
Phospho-p38 MAPK (Thr179+Tyr181) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p38 MAPK (Thr179+Tyr181) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q16539. Gene ID: 1432. Target Synonyms: protein; kinase p38-alpha; MAP kinase MXI2; MAP kinase p38; MAPK14; MAX-interacting protein 2; Mitogen-activated protein kinase 14; Mitogen-activated protein kinase p38; MK14; MXI2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.