Learn More
Biomatik Corporation Phospho-MDM2 (Ser166) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51122
Encompass_Preferred
Phospho-MDM2 (Ser166) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MDM2 (Ser166) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q00987. Gene ID: 4193. Target Synonyms: Double minute 2 protein; HDM2; Oncoprotein Mdm2; p53-binding protein Mdm2; Ubiquitin-protein ligase E3 Mdm2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9016
Explore available promotions