Learn More
Biomatik Corporation Phospho-CSFR (Tyr561) Colorimetric Cell-Based ELISA Kit
Supplier: Biomatik Corporation EKA51279
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Phospho-CSFR (Tyr561) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CSFR (Tyr561) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P07333. Gene ID: 1436. Target Synonyms: c-fms; CD115; CSF-1-R; CSF1R; CSFMR; FMS; Fms proto-oncogene; kinase CSFR; M-CSFR; Macrophage colony stimulating factor I receptor precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.