Learn More
SPS Medical ECONOMY MONITORING SYSTEM
Supplier: SPS Medical EMS012
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Our value system provides customers with one test strip and one control strip which meets all local and national guidelines for spore testing. Steam test results are documented after 24 hours incubation in our laboratory. Test reports are sent on a quarterly basis, though access to spore test results are available any time at www.checkyourtest.com. The EMS is also certified for use in Dry Heat, Chemical Vapor, and EO Gas processes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.