Learn More
Across International ECO 150C 0.9 Cu Ft Vacuum Drying Oven with LED Lights
SDP
Supplier:  Across International AT09E
Encompass_Preferred
AT09e series digital desktop vacuum ovens feature a 0.9 cubic foot easy-to-clean stainless steel chamber with a large dual-layer tempered glass safety window and small footprint. These ovens can greatly reduce drying time by maintaining a consistent vacuum level within the chamber. Every one of our vacuum ovens goes through a 2-time 24-hour vacuum leak test, and is 100% quality controlled in New Jersey or Nevada before leaving our facilities.
Catalog No. 50-225-1183
Explore available promotions