Learn More
Bioworld ECH SEPAROPOREAGAROSE) 4B
Supplier: Bioworld 201810083
ECH Separopore 4B Affinity matrix for stable carbodiimide-based coupling of ligands containing amino groups.; Ligands are to be coupled in a single-step procedure in the presence of a coupling reagent (not provided), a water-soluble carb...