Learn More
Bioworld ECH SEPAROPOREAGAROSE) 4B
Supplier:  Bioworld 201810081
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements. ECH Separopore 4B Affinity matrix for stable carbodiimide-based coupling of ligands containing amino groups.; Ligands are to be coupled in a single-step procedure in the presence of a coupling reagent (not provided), a water-soluble carb...
Catalog No. 50-197-675
Explore available promotions