Learn More
Perkin Elmer US LLC D-Torch w/1-Slot, Assembly w/Quartz Outer Tube for Optima 8x00
Supplier: Perkin Elmer US LLC N0777840
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The D-Torch is a revolutionary new demountable torch design with interchangeable outer tubes. Featuring a torch body with fixed alumina intermediate tube for greater robustness, in addition to a demountable outer tube and injector, the D-Torch significantly reduces torch replacement costs for the analysis of challenging sample matrices. The quartz outer tube is ideal for most aqueous samples, while the ceramic outer tube may last longer for samples with high dissolving solids or for organic applications, such as wear metals in engine oils. The D-Torch also features an O-ring free torch body for simplified maintenance and is compatible with all commonly used spray chambers.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.