Learn More
Perkin Elmer US LLC D-Torch Complete Assembly with Quartz Outer Tube for Avio 200
Supplier: Perkin Elmer US LLC N0790442
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The D-Torch provides a demountable torch design without sacrificing performance or usability. Featuring a torch body with fixed alumina intermediate tube for greater robustness, in addition to a demountable outer tube and injector, the D-Torch significantly reduces torch replacement costs for the analysis of challenging sample matrices. Includes: N0791300 D Torch Body Avio Assembly 31-808-3802 N0791301 D-Torch PE AVIO Quartz Outer Tube Assy (Single Slot) 31-808-3792 N0791302 Retaining Ring for D-Torch Outer Tube 31-808-2818 N0777421-med Safety Gloves Medium SAFEGLOVE-med
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.