Learn More
Perkin Elmer US LLC D-Torch Complete Assembly with Quartz Outer Tube for Avio 500
Supplier: Perkin Elmer US LLC N0810861
Features and Benefits include: An alumina intermediate tube which resists wear and is tolerant to high temperatures Demountable outer tube With the D-Torch, you need only replace the outer tube Interchangeable quartz and ceramic outer tubes Interchangeable injectors choose the optimum injector for your application. Select from quartz for aqueous solutions, narrow bore for organics, wide bore for high dissolved solids or ceramic for HF O-ring free torch body for simplified maintenance and improved reliability
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.