Learn More
Chemglass Life Sciences Drying Tube, U Shaped, 14/20 Inner Joint, # 00 Rubber Stopper
Supplier: Chemglass Life Sciences CG129614
U shaped drying tube supplied complete with rubber stopper, glass inlet tube, and a lower inner joint.