Learn More
United States Biological Corporation Drop-out Mix Synthetic, Adenine Rich (2g) Minus Tryptophan w/o Yeast Nitrogen Base (DO, Dropout, Powder) | 100g
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: United States Biological Corporation D9531100G
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Drop-out Mixes are used as a supplement to Drop-out Base for making synthetic, complete media formulations in accordance with Methods in Yeast Genetics. All lots are quality control tested using the appropriate yeast strains and transformants.