Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Urine Drugs of Abuse Calibrators

Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with these ready-to-use liquid assays.

$129.00

Specifications

Content And Storage 2°C to 8°C
Concentration Multianalyte
Detectable Analytes Amphetamine, Cocaine, Opiate, Phencyclidine (PCP)
Sample Type Urine
Type Calibrator
View More Specs

Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
23-278-298
View Documents
Thermo Scientific™
0034
DRI; Low Calibrator; 5mL
Each for $129.00
Add to Cart
 
23-278-299
View Documents
Thermo Scientific™
0036
DRI; High Calibrator; 5mL
Each for $129.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Urine Drugs of Abuse Calibrators—ready-to-use assays in liquid form. These assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
Specifications

Specifications

2°C to 8°C
Amphetamine, Cocaine, Opiate, Phencyclidine (PCP)
Calibrator
1 x 5 mL
Multianalyte
Urine
1 Bottle
SDS
Documents

Documents

Product Certifications
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.