Learn More
Thermo Scientific™ DRI™ Benzodiazepine Drugs of Abuse Assays
Save time in reagent preparation while maintaining high analytical accuracy with Thermo Scientific™ DRI™ Benzodiazepine Drugs of Abuse Assays.
$1347.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Control Sets | MGC Primary DAU Control Set |
Detectable Analytes | Oxazepam |
DoA Calibrators | DRI Multi-drug Calibrators |
Sample Type | Urine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-278-301
|
Thermo Scientific™
0039 |
1 x 100 mL |
Each for $2,606.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-356
|
Thermo Scientific™
10015644 |
3 x 18 mL |
Each for $1,347.00
|
|
|||||
Description
Save time in reagent preparation while maintaining high analytical accuracy with Thermo Scientific™ DRI™ Benzodiazepine Drugs of Abuse Assays. These liquid, ready-to-use assays offer applications for an array of clinical chemistry analyzers and have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Variety of calibrators and multiconstituent control sets
Specifications
2°C to 8°C | |
Oxazepam | |
Urine |
MGC Primary DAU Control Set | |
DRI Multi-drug Calibrators | |
Reagent |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.