Learn More
Mystaire™ Downflow Workstation Main Filter
For use with the Mystaire™ Downflow Workstation (FE-WS6). Mystaire™ filters are manufactured to the strictest quality standards in the industry to ensure their capability to meet your application challenges.
$736.00 - $934.00
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity & Availability | |||||
15-338-193
|
Mystaire™
FE5007 |
For Formaldehyde- FE-WS6 |
Each for $934.00
|
|
|||||
15-338-192
|
Mystaire™
FE5004 |
For Hydrocarbons- FE-WS6 |
Each for $736.00
|
|
|||||
15-338-194
|
Mystaire™
FE5014 |
For Mineral Acids- FE-WS6 |
Each for $772.00
|
|
|||||
Description
- Proper filtration selection is critical to ensure operator safety from potentially harmful vapors and airborne particulate.
- Made in the U.S.A.
- Each filter application is reviewed to ensure a proper filtration package is selected
- Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)
Specifications
FE-WS6 | |
Carbon | |
Filter |