Learn More
Clontech Labs 3P DNA-OFF™, 500 mL
SDP
Supplier:  Clontech Labs 3P 9036
Encompass
DNA-OFF is a non-alkaline, non-corrosive and non-carcinogenic cleansing solution to eliminate DNA contamination and is intended for use at PCR workstations. Contamination of DNA at a PCR work area can result in amplification artifacts. The solution contains a surfactant plus an agent that destroys DNA. DNA-OFF is stable, heat resistant, and ready-to-use. 500 mL
Catalog No. 50-443-878
Explore available promotions