Learn More
Enzo Life Sciences 1,2-Dilauroyl-sn-glycero-3-phosphocholine (250mg). CAS: 18194-25-7
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Enzo Life Sciences ALX300034M250
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Alternative name: DLPC. Purity: ≥99% (TLC). Appearance: White to off-white solid. Solubility: 20mg/ml soluble in chloroform:methanol:acetic acid:water (100:30:10:2.5); slightly soluble in methanol. Long Term Storage: -20°C.