Learn More
CTL Scientific Supply Corp DIPYRIDYL PAPER,200 STRIPS
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 90725
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
DIPYRIDYL PAPER - 200 strips. The test paper allows the specific detection of Fe(II) ions. Even small amounts of Fe(II) can safely be detected in presence of large amounts of Fe(III). Limit of sensitivity: 2 mg/l Fe 2+