Learn More
Quantimetrix DipperPOCT UA DIPSTICK CONTROL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Quantimetrix 140001
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
• Three months of room temperature stabilityand up to 3 years at 2ºC–8ºC. • Full dipstick immersion. • Zero wasted QC product