Learn More
Medchemexpress LLC HY-128454 100mg Medchemexpress, Dimethyl trisulfide CAS:3658-80-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128454100MG
Encompass_Preferred
Medchemexpress, HY-128454 100mg Dimethyl trisulfide CAS:3658-80-8 Dimethyl trisulfide is an organic chemical compound and the simplest organic trisulfide found in garlic, onion, broccoli, and similar plants. Dimethyl trisulfide is a cyanide antidote. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9750
Explore available promotions