Learn More
Siemens Healthineers Dimertest Latex Assay Beads
For use with DIMERTEST™ Latex kit for the rapid qualitative or semiquantitative evaluation of circulating derivatives of cross-linked fibrin degradation products (XL-FDP) in human plasma.
Supplier: Siemens Healthineers 10445723
This item is not returnable.
View return policy
Description
DIMERTEST™ Latex kits are rapid agglutination assays that use latex particles coated with a specific D-dimer monoclonal antibody. It is intended for the qualitative or semiquantitative evaluation of cross-linked fibrin degradation products containing D-dimers.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Hemostasis Reagent | |
D-Dimer Testing | |
1 x 2 mL Beads |