Learn More
BD Difco™ Poly Antisera
For agglutination test
Supplier: BD 228341
This item is not returnable.
View return policy
Description
Salmonella poly antiserum
- Febrile control antisera
- Helps in the detection of diseases such as salmonellosis, brucellosis and rickettsial diseases
- Qualitative and semi-quantitative slide or tube agglutination tests for homologous antibodies
- Used along with febrile antigens as positive or negative controls
- For identification of morphologically and biochemically identified Shigella species
- Can be used only in slide agglutination test
- Lyophilized, polyclonal rabbit antisera
Specifications
For in vitro diagnostic use | |
Shigella | |
3 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For in vitro diagnostic use