Learn More
Diamedix™ Immunosimplicity™ Rheumatoid Factor Test Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Detects RF IgM-class antibodies in human serum
Supplier: Diamedix™ 720710
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Alert:
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes: Antigen Wells, Cut-off Calibrator, Postive Control, Negative Control, Sample E Diluent, Wash U Concentrate (20X), Conjugate, Substrate HRP, Stop O Solution
Description
The Diamedix™ Immunosimplicity™ Rheumatoid Factor Test Kit is a quantitative indirect enzyme immunoassay
- Intended for the quantitation of RF IgM antibodies in human serum
- Aids in the diagnosis of rheumatoid arthritis (RA)
- For in vitro diagnostic use
- Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
Specifications
Rheumatoid Factor (RF) IgM | |
Immunosimplicity™ Rheumatoid Factor Test Kit | |
Antigen wells, cut-off calibrator, positive control, negative control, sample E Diluent, wash U concentrate 20X, conjugate, substrate HRP, stop O solution | |
96 Tests |