Learn More
Enzo Life Sciences Dengue NS1 ST2, (recombinant) (100µg)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Enzo Life Sciences ENZPRT1050100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Alternative name: DENV1-NS1. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In DPBS pH 7.4. Source: Produced in HEK 293 cells. Dengue Viral NS1 Serotype 2 (DENV2-NS1) from strain Thailand/16681/84. Long Term Storage: -80°C. Applications: ELISA Vaccine research, LINE assay, Antibody capture.