Learn More
Nelson Jameson Company Delvocid XT1, 500 g
Supplier: Nelson Jameson Company 2334060
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Delvocid XT1 is a powdered form with a xanthan gum stabilizer which provides a stable suspension to eliminate the need for constant stirring of the preparation. Delvocid is FDA accepted for use as a cheese preservative when used properly. Kosher-approved.