Learn More
Ampco Safety Tools Deep Well Socket, 1/4 in. Drive (Imperial)
SDP
AMPCO™ Deep Well Socket performs in high-risk and demanding environments.
$182.00 - $207.00
Specifications
Product Type Deep-well Socket Wrench
Drive Size 1/4 in.
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
View More Specs
Alert:
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Products 10
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
19-173-653
SDP
View Documents
Ampco Safety Tools
DW1/4D1/2
1/2 in.
Each for $197.00
Add to Cart
19-173-654
SDP
View Documents
Ampco Safety Tools
DW1/4D1/4
1/4 in.
Each for $182.00
Add to Cart
19-173-656
SDP
View Documents
Ampco Safety Tools
DW1/4D11/32
11/32 in.
Each for $185.00
Add to Cart
19-173-661
SDP
View Documents
Ampco Safety Tools
DW1/4D3/16
3/16 in.
Each for $182.00
Add to Cart
19-173-662
SDP
View Documents
Ampco Safety Tools
DW1/4D3/8
3/8 in.
Each for $189.00
Add to Cart
19-173-666
SDP
View Documents
Ampco Safety Tools
DW1/4D5/16
5/16 in.
Each for $182.00
Add to Cart
19-173-669
SDP
View Documents
Ampco Safety Tools
DW1/4D7/16
7/16 in.
Each for $197.00
Add to Cart
19-173-670
SDP
View Documents
Ampco Safety Tools
DW1/4D7/32
7/32 in.
Each for $182.00
Add to Cart
19-173-673
SDP
View Documents
Ampco Safety Tools
DW1/4D9/16
9/16 in.
Each for $207.00
Add to Cart
19-173-674
SDP
View Documents
Ampco Safety Tools
DW1/4D9/32
9/32 in.
Each for $182.00
Add to Cart
Description
Description
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Specifications
Deep-well Socket Wrench
1/4 in.
Aluminum Bronze
Factory Mutual (FM) Approved
SDS
Documents
Documents
Product Certifications
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.