Learn More
Revvity Health Sciences Inc D-dimer (human), lyophilized (AlphaLISA)
Supplier: Revvity Health Sciences Inc AL290S
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
This standard should be reconstituted in 100 ¿L Milli-Q grade water. The reconstituted analyte should be used within 60 minutes or aliquoted into screw-capped polypropylene vials and stored at -20¿C for further experiments. Avoid multiple freeze-thaw cycles. One vial contains an amount of analyte sufficient for performing 10 standard curves.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.