Learn More
Electron Microscopy Sciences Double Slot Set Screw Vise, 15 mm
Supplier: Electron Microscopy Sciences 7595103
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
SEM specimen holder with two 1 mm by 3 mm deep slots. Clamp thin specimens and cross sections without conductive paint or tape. 3.2 mm pin. Machined aluminum with 2 stainless steel allen set screws. Includes allen wrench.