Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ DBI-SALA™ Talon™ Twin-Leg Tie-Back Quick Connect Self Retracting Lifeline
Provides continuous fall protection and significant increase in mobility.
Supplier: 3M™ 3102101
Description
- Twin-leg 100% tie-off design
- Built-in connector for direct attachment to harness
- Delta™ comfort pad and lanyard keepers
- Compact, lightweight design
- Tough and impact resistant thermoplastic housing
- Inertia activated brake system
- Self-locking steel rebar hooks
- Impact indicator
- i-Safe™ equipped
- 2.3m (7.5 ft.) 2.5cm (1 in.) nylon web tie-back lifelines
Specifications
OSHA 1910.66, OSHA 1926.502 | |
159 kg | |
Construction, General Industrial, Oil and Gas, Transportation, Utilities |
2.3 m | |
7.5 ft. | |
350 lb. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction