Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-SALA™ Advanced™ One-Piece Adjustable Offset Davit Mast

Variable offset davits are designed for use where multiple offset masts are required. For applications with overhead clearance restrictions, the adjustable height of the variable offset masts can eliminate the need for custom length masts and mast extensions. Quick connect/release pins allow for fast and easy set-up, dismantle and transport.

$5722.50 - $7591.50

Specifications

For Use With (Application) Construction, General Industrial, Mining, Oil and Gas, Transportation, Utilities
Includes One-Piece Adjustable Offset Davit Mast
Strength 5,000 lb. (22 kN) mast anchorage point
View More Specs

Products 4
Catalog Number Mfr. No. Certifications/Compliance Product Type Weight (English) Price Quantity & Availability  
Catalog Number Mfr. No. Certifications/Compliance Product Type Weight (English) Price Quantity & Availability  
19-070-829
View Documents
3M™
7100204340
ANSI Z359.1, OSHA 1926.502 66 in. to 76 in. (167.6 to 193 cm) anchor point height 31 lb.
Case for $5,722.50
Add to Cart
 
19-070-827
View Documents
3M™
7100204341
ANSI A10.32, ANSI Z359.1, OSHA 1926.502 66 in. to 76 in. (167.6 to 193 cm) anchor point height 31.1 lb.
Case for $5,764.50
Add to Cart
 
19-070-830
View Documents
3M™
7100204337
OSHA 1926.502, OSHA 1910.66 69.5 in. to 87.5 in. (176.5 to 222.2 cm) anchor point height 40 lb.
Case for $6,751.50
Add to Cart
 
19-070-820
View Documents
3M™
7100204332
OSHA 1910.66, OSHA 1926.502 93.5 in. to 111.5 in. (237.5 to 282.2 cm) anchor point height 50 lb.
Case for $7,591.50
Add to Cart
 
Description

Description

  • Portable install
  • Fall arrest rated in all davit offset mast positions
  • Integral secondary fall arrest anchorage point
  • Built-in primary and secondary pulleys
  • Quick mount winch/SRL bracket
  • Configure with front and/or back mounted winch/SRL
  • Detent and knob style adjustment
  • Lightweight, corrosion resistant aluminum construction
  • Use with Advanced™ portable and permanent bases
  • Specifications

    Specifications

    Construction, General Industrial, Mining, Oil and Gas, Transportation, Utilities
    One-Piece Adjustable Offset Davit Mast
    5,000 lb. (22 kN) mast anchorage point
    SDS
    Documents

    Documents

    Product Certifications
    Provide Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit