Learn More
United States Biological Corporation D+ GLUCOSE ANHYDROUS
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: United States Biological Corporation G305010KG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Used as a carbohydrate source in cell culture and molecular biology. Appearance: White, crystalline powder Purity: ≥99.5% Identity: To pass USP Specific Rotation: +52.6° to +53.2° Loss on Drying: ≤0.2% Residue on Ignition: ≤0.1% Acidity, ml: To pass USP Starch: To pass USP Dextrin: To pass USP Chloride: ≤0.018% Sulfate: ≤0.025% Arsenic: ≤0.0001% Heavy Metals (Pb): ≤ 0.0005% Meets or exceeds USP specifications Certificate of Origin: USA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.