Learn More
Techcyte Cytobay Bladder Cancer Analysis
Cytobay Bladder Cancer Analysis, combined with CytoBay’s proprietary stain, automatically detects ICC positive low grade, high grade, and other diagnostically significant cells.
Supplier: Techcyte BCPAYG
Description
- Software that automates cell locating from a digital image of a human urine sample intended for in-vitro diagnostic use
- Detects the presence or absence of bladder cancer cells.
- The user (technologist or pathologist) is presented with a viewer that provides pre-classified cells of ICC Positive (high and low grade urothelial carcinoma), Uroepithelial, and Normal Squamous
- Enables the user to give final confirmation of the presence or absence of bladder cancer cells
- Intended to be used in conjunction with complete standard of care evaluation of whole slide images
Specifications
Detection Software | |
Microsoft Windows 10 IoT 64-bit | |
Analyzing Data |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For investigational use only in the US.