Learn More
PERFECTOR SCIENTIFIC Cuvette Caps 500/pk
Supplier: PERFECTOR SCIENTIFIC 9020
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Polyethylene cap is liquid tight but very easy to insert and remove making the cuvettes suitable for mixing and storage. 500/pk