Learn More
Fireflysci SEPTA for open screw caps (SC2)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Fireflysci SEPTA
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Black screw cap with 13-425 threading (comes with PTFE septa)• Is an open cap that enables injection of sample through a self-healing injection site