Learn More
Sartorius Cubis™ II Precision (1-place) Balance, MCE Interface
Cubis™ II Precision (1-Place) Balance features fully customizable hardware, software, and connectivity
$6279.00 - $13041.00
Specifications
Connection Type 2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Display Type TFT Touchscreen
Material Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
Type Precision (1-place) Balance
Warranty 10 years
View More Specs
Products 7
Catalog Number Mfr. No. Capacity Model Includes Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Model Includes Price Quantity & Availability  
14-561-213
View Documents Promotion Details
Sartorius
MCE5202S2S00A
5200 g Draft shield A - Motorized Balance, Accessories, Power supply, Manual, Factory test report
Each for $11,502.00
Add to Cart
14-561-214
View Documents Promotion Details
Sartorius
MCE5202S2S00E
5200 g Draft shield E - Small glass Balance, Accessories, Power supply, Manual, Factory test report
Each for $7,765.00
Add to Cart
14-561-215
View Documents Promotion Details
Sartorius
MCE5202S2S00I
5200 g Draft shield I - Motorized with ionizer Balance, Accessories, Power supply, Manual, Factory test report
Each for $13,041.00
Add to Cart
14-561-216
View Documents Promotion Details
Sartorius
MCE5202S2S00R
5200 g Draft shield R - Stainless steel frame Balance, Accessories, Power supply, Manual, Factory test report
Each for $7,765.00
Add to Cart
14-561-217
View Documents Promotion Details
Sartorius
MCE5202S2S00U
5200 g Draft shield U - Manual glass Balance, Accessories, Power supply, Manual, Factory test report
Each for $8,383.00
Add to Cart
14-561-277
View Documents Promotion Details
Sartorius
MCE8201S0S00
8200 g No draft shield Balance, Accessories, Power supply, Manual, Factory test report, QP99 Software
Each for $6,317.00
Add to Cart
14-561-260
View Documents Promotion Details
Sartorius
MCE12201S0S00
12,200 g No draft shield Balance, Accessories, Power supply, Manual, Factory test report, QP99 Software
Each for $6,279.00
Add to Cart
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Warranty and Services
10-year warranty
Specifications
Specifications
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
10 years
5.4 kg
100/240 V
TFT Touchscreen
Precision (1-place) Balance
240 mm
120/240 V
50/60 Hz (± 5%)