Learn More
Thermo Scientific Thermal Cutoff Kit 1PK
Supplier: Thermo Scientific 088662
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
600 MM long anion trap column containing polymethacrylate based anion exchange resin in hydroxide form designed to remove trace chloride, sulfate, carbonate anions from eluent prior to entering IC separation column, thereby improving baseline stability & noise, has 4 MM internal diameter & is made of inert polyether ether ketone polymer, can be used with common IC eluents like sodium hydroxide & sodium carbonate solutions.
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.